Assay Details
| Gene Symbol: spsb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025460.1 | NP_001020631.1 | 2 - 3 | 1021 | 74 | |
| XM_021469752.1 | XP_021325427.1 | 2 - 3 | 928 | 74 | ||
| GenBank mRNA | BC097174.1 | - | 2 - 3 | 1021 | 74 | |
More Information
Set Membership: |
|

spsb1
Genomic Map