Assay Details
| Gene Symbol: paqr3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030148.2 | NP_001025319.2 | 3 - 4 | 531 | 101 | |
| XM_005155617.4 | XP_005155674.1 | 3 - 4 | 514 | 101 | ||
| XM_005155618.4 | XP_005155675.1 | 3 - 4 | 516 | 101 | ||
| GenBank mRNA | BC096792.1 | - | 3 - 4 | 101 | ||
More Information
Set Membership: |
|

paqr3b
Genomic Map