Assay Details
| Gene Symbol: necab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328213.1 | NP_001315142.1 | 6 - 7 | 725 | 66 | |
| XM_009293572.3 | XP_009291847.1 | 6 - 7 | 760 | 66 | ||
| GenBank mRNA | BC098611.1 | - | 6 - 7 | 707 | 66 | |
More Information
Set Membership: |
|

necab2
Genomic Map