Assay Details
| Gene Symbol: srsf7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030267.1 | NP_001025438.1 | 463 | 74 | ||
| GenBank mRNA | BC097250.1 | - | 463 | 74 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

srsf7b| Gene Symbol: srsf7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030267.1 | NP_001025438.1 | 463 | 74 | ||
| GenBank mRNA | BC097250.1 | - | 463 | 74 | ||
Set Membership: |
|