Assay Details
| Gene Symbol: pabpc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031676.1 | NP_001026846.1 | 7 - 8 | 1285 | 70 | |
| XM_005169771.4 | XP_005169828.1 | 7 - 8 | 1315 | 70 | ||
| GenBank mRNA | BC047811.1 | - | 7 - 8 | 1004 | 70 | |
| BC099992.1 | - | 7 - 8 | 1285 | 70 | ||
More Information
Set Membership: |
|

pabpc1a
Genomic Map