Assay Details
| Gene Symbol: adal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033744.1 | NP_001028916.1 | 1 - 2 | 99 | 92 | |
| GenBank mRNA | BC096787.1 | - | 1 - 2 | 99 | 92 | |
| BC152246.1 | - | 1 - 2 | 147 | 92 | ||
More Information
Set Membership: |
|

adal
Genomic Map