Assay Details
| Gene Symbol: slit1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034975.1 | NP_001030147.1 | 26 - 27 | 2813 | 60 | |
| XM_009296502.3 | XP_009294777.1 | 26 - 27 | 2934 | 60 | ||
| XM_021469336.1 | XP_021325011.1 | 26 - 27 | 2940 | 60 | ||
| GenBank mRNA | BC163568.1 | - | 26 - 27 | 2801 | 60 | |
| CU638802.1 | - | 26 - 27 | 2826 | 60 | ||
| DQ153065.1 | - | 26 - 27 | 2813 | 60 | ||
More Information
Set Membership: |
|

slit1b
Genomic Map