Assay Details
| Gene Symbol: slit1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034975.1 | NP_001030147.1 | 32 - 33 | 3632 | 107 | |
| XM_009296502.3 | XP_009294777.1 | 32 - 33 | 3753 | 107 | ||
| XM_021469336.1 | XP_021325011.1 | 32 - 33 | 3759 | 107 | ||
| GenBank mRNA | CU638802.1 | - | 32 - 33 | 3645 | 107 | |
| DQ153065.1 | - | 32 - 33 | 3632 | 107 | ||
More Information
Set Membership: |
|

slit1b
Genomic Map