Assay Details
| Gene Symbol: mmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037419.1 | NP_001032496.1 | 3 - 4 | 522 | 133 | |
| GenBank mRNA | BC109439.1 | - | 3 - 4 | 522 | 133 | |
More Information
Set Membership: |
|

mmd
Genomic Map