Assay Details
| Gene Symbol: sspn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037580.1 | NP_001032669.1 | 1 - 2 | 367 | 61 | |
| GenBank mRNA | BC110092.1 | - | 1 - 2 | 367 | 61 | |
More Information
Set Membership: |
|

sspn
Genomic Map