Assay Details
| Gene Symbol: wt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039634.2 | NP_001034723.2 | 3 - 4 | 760 | 74 | |
| XM_005159243.4 | XP_005159300.1 | 3 - 4 | 74 | |||
| GenBank mRNA | BC124087.1 | - | 3 - 4 | 760 | 74 | |
| BC164718.1 | - | 3 - 4 | 618 | 74 | ||
| DQ145942.1 | - | 3 - 4 | 74 | |||
More Information
Set Membership: |
|

wt1b
Genomic Map