Assay Details
| Gene Symbol: wt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039634.2 | NP_001034723.2 | 7 - 8 | 1175 | 71 | |
| XM_005159243.4 | XP_005159300.1 | 7 - 8 | 1448 | 71 | ||
| GenBank mRNA | BC124087.1 | - | 7 - 8 | 1175 | 71 | |
| BC164718.1 | - | 7 - 8 | 1033 | 71 | ||
| DQ145942.1 | - | 7 - 8 | 999 | 71 | ||
More Information
Set Membership: |
|

wt1b
Genomic Map