Assay Details
| Gene Symbol: wt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039634.2 | NP_001034723.2 | 8 - 9 | 1272 | 54 | |
| GenBank mRNA | BC124087.1 | - | 8 - 9 | 1272 | 54 | |
| BC164718.1 | - | 8 - 9 | 1130 | 54 | ||
| DQ145942.1 | - | 8 - 9 | 1096 | 54 | ||
More Information
Set Membership: |
|

wt1b
Genomic Map