Assay Details
| Gene Symbol: specc1la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039816.1 | NP_001034905.1 | 10 - 11 | 2830 | 63 | |
| XM_005167177.4 | XP_005167234.1 | 11 - 12 | 3022 | 63 | ||
| XM_009304055.3 | XP_009302330.1 | 11 - 12 | 3022 | 63 | ||
| GenBank mRNA | AY884302.1 | - | 10 - 11 | 2830 | 63 | |
More Information
Set Membership: |
|

specc1la
Genomic Map