Assay Details
| Gene Symbol: rpl31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040052.2 | NP_001035141.1 | 2 - 3 | 127 | 117 | |
| GenBank mRNA | BC114301.1 | - | 106 | 117 | ||
| EH441458.1 | - | 2 - 3 | 156 | 117 | ||
| EH539710.1 | - | 2 - 3 | 166 | 117 | ||
More Information
Set Membership: |
|

rpl31
Genomic Map