Assay Details
| Gene Symbol: rpl31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040052.2 | NP_001035141.1 | 4 - 5 | 360 | 148 | |
| GenBank mRNA | BC114301.1 | - | 339 | 148 | ||
More Information
Set Membership: |
|

rpl31
Genomic Map