Assay Details
| Gene Symbol: dab1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040386.1 | NP_001035476.1 | 2 - 3 | 474 | 136 | |
| NM_001135042.1 | NP_001128514.1 | 2 - 3 | 474 | 136 | ||
| XM_005171112.4 | XP_005171169.1 | 2 - 3 | 474 | 136 | ||
| XM_005171115.4 | XP_005171172.1 | 2 - 3 | 474 | 136 | ||
| GenBank mRNA | BC115171.1 | - | 2 - 3 | 474 | 136 | |
More Information
Set Membership: |
|

dab1b
Genomic Map