Assay Details
| Gene Symbol: dab1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040386.1 | NP_001035476.1 | 6 - 7 | 955 | 128 | |
| NM_001135042.1 | NP_001128514.1 | 6 - 7 | 955 | 128 | ||
| XM_005171110.4 | XP_005171167.1 | 6 - 7 | 1145 | 128 | ||
| XM_005171111.4 | XP_005171168.1 | 6 - 7 | 1145 | 128 | ||
| XM_005171112.4 | XP_005171169.1 | 6 - 7 | 955 | 128 | ||
| XM_005171114.4 | XP_005171171.1 | 6 - 7 | 1145 | 128 | ||
| XM_005171115.4 | XP_005171172.1 | 6 - 7 | 955 | 128 | ||
| XM_005171116.4 | XP_005171173.1 | 6 - 7 | 1146 | 128 | ||
| XM_021481242.1 | XP_021336917.1 | 6 - 7 | 1145 | 128 | ||
| GenBank mRNA | BC115171.1 | - | 6 - 7 | 955 | 128 | |
| DQ883567.1 | - | 5 - 6 | 622 | 128 | ||
| EF028079.1 | - | 5 - 6 | 622 | 128 | ||
More Information
Set Membership: |
|

dab1b
Genomic Map