Assay Details
| Gene Symbol: c6ast1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043319.1 | NP_001036784.1 | 1 - 2 | 62 | 76 | |
| GenBank mRNA | AB256952.1 | - | 1 - 2 | 62 | 76 | |
More Information
Set Membership: |
|

c6ast1
Genomic Map