Assay Details
| Gene Symbol: c6ast1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043319.1 | NP_001036784.1 | 4 - 5 | 391 | 101 | |
| GenBank mRNA | AB256952.1 | - | 4 - 5 | 391 | 101 | |
| BC162740.1 | - | 5 - 6 | 459 | 101 | ||
More Information
Set Membership: |
|

c6ast1
Genomic Map