Assay Details
| Gene Symbol: bloc1s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044350.2 | NP_001037815.1 | 3 - 4 | 337 | 70 | |
| GenBank mRNA | BC135064.1 | - | 3 - 4 | 337 | 70 | |
More Information
Set Membership: |
|

bloc1s1
Genomic Map