Assay Details
| Gene Symbol: ighmbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044900.1 | NP_001038365.1 | 13 - 14 | 2705 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ighmbp2| Gene Symbol: ighmbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044900.1 | NP_001038365.1 | 13 - 14 | 2705 | 104 | |
Set Membership: |
|