Assay Details
| Gene Symbol: ptpn23b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044928.1 | NP_001038393.1 | 1 - 2 | 261 | 96 | |
| XM_005159517.4 | XP_005159574.1 | 1 - 2 | 292 | 96 | ||
| GenBank mRNA | BC171436.1 | - | 1 - 2 | 213 | 96 | |
More Information
Set Membership: |
|

ptpn23b
Genomic Map