Assay Details
| Gene Symbol: ptpn23b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044928.1 | NP_001038393.1 | 2 - 3 | 331 | 77 | |
| XM_005159517.4 | XP_005159574.1 | 2 - 3 | 362 | 77 | ||
| GenBank mRNA | BC171436.1 | - | 2 - 3 | 283 | 77 | |
More Information
Set Membership: |
|

ptpn23b
Genomic Map