Assay Details
| Gene Symbol: ptpn23b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044928.1 | NP_001038393.1 | 3 - 4 | 459 | 82 | |
| XM_005159517.4 | XP_005159574.1 | 3 - 4 | 490 | 82 | ||
| GenBank mRNA | BC171436.1 | - | 3 - 4 | 411 | 82 | |
More Information
Set Membership: |
|

ptpn23b
Genomic Map