Assay Details
| Gene Symbol: ptpn23b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044928.1 | NP_001038393.1 | 7 - 8 | 797 | 121 | |
| XM_005159517.4 | XP_005159574.1 | 7 - 8 | 828 | 121 | ||
| GenBank mRNA | BC171436.1 | - | 7 - 8 | 749 | 121 | |
| FN428734.1 | - | 4 - 5 | 322 | 121 | ||
More Information
Set Membership: |
|

ptpn23b
Genomic Map