Assay Details
| Gene Symbol: slc12a10.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045001.1 | NP_001038466.1 | 1 - 2 | 251 | 77 | |
| GenBank mRNA | EF591989.1 | - | 1 - 2 | 233 | 77 | |
More Information
Set Membership: |
|

slc12a10.2
Genomic Map