Assay Details
| Gene Symbol: slc12a10.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045001.1 | NP_001038466.1 | 24 - 25 | 2797 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

slc12a10.2| Gene Symbol: slc12a10.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045001.1 | NP_001038466.1 | 24 - 25 | 2797 | 83 | |
Set Membership: |
|