Assay Details
| Gene Symbol: ripor2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045026.1 | NP_001038491.1 | 11 - 12 | 1125 | 101 | |
| XM_005159634.4 | XP_005159691.1 | 11 - 12 | 1172 | 101 | ||
| XM_009294297.3 | XP_009292572.1 | 11 - 12 | 1039 | 101 | ||
| XM_021468090.1 | XP_021323765.1 | 11 - 12 | 1287 | 101 | ||
| XM_021468091.1 | XP_021323766.1 | 11 - 12 | 1195 | 101 | ||
More Information
Set Membership: |
|

ripor2
Genomic Map