Assay Details
| Gene Symbol: ripor2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045026.1 | NP_001038491.1 | 12 - 13 | 1253 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ripor2| Gene Symbol: ripor2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045026.1 | NP_001038491.1 | 12 - 13 | 1253 | 92 | |
Set Membership: |
|