Assay Details
| Gene Symbol: tfe3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045066.1 | NP_001038531.1 | 4 - 5 | 648 | 85 | |
| XM_005169305.4 | XP_005169362.1 | 5 - 6 | 766 | 85 | ||
| XM_017358245.2 | XP_017213734.1 | 2 - 3 | 565 | 85 | ||
| GenBank mRNA | JN105117.1 | - | 4 - 5 | 648 | 85 | |
More Information
Set Membership: |
|

tfe3b
Genomic Map