Assay Details
| Gene Symbol: tfe3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045066.1 | NP_001038531.1 | 6 - 7 | 777 | 107 | |
| XM_005169305.4 | XP_005169362.1 | 7 - 8 | 895 | 107 | ||
| XM_005169306.4 | XP_005169363.1 | 7 - 8 | 889 | 107 | ||
| XM_005169307.4 | XP_005169364.1 | 7 - 8 | 877 | 107 | ||
| XM_005169308.4 | XP_005169365.1 | 7 - 8 | 871 | 107 | ||
| XM_009306036.3 | XP_009304311.1 | 7 - 8 | 883 | 107 | ||
| XM_017358245.2 | XP_017213734.1 | 4 - 5 | 694 | 107 | ||
| GenBank mRNA | JN105115.1 | - | 6 - 7 | 768 | 107 | |
More Information
Set Membership: |
|

tfe3b
Genomic Map