Assay Details
| Gene Symbol: ahsa1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045091.1 | NP_001038556.1 | 5 - 6 | 826 | 130 | |
| XM_005160576.4 | XP_005160633.1 | 5 - 6 | 1126 | 130 | ||
| GenBank mRNA | BC114279.1 | - | 5 - 6 | 834 | 130 | |
More Information
Set Membership: |
|

ahsa1a
Genomic Map