Assay Details
| Gene Symbol: lgalsla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045300.1 | NP_001038765.1 | 4 - 5 | 454 | 79 | |
| GenBank mRNA | BC117644.1 | - | 4 - 5 | 454 | 79 | |
More Information
Set Membership: |
|

lgalsla
Genomic Map