Assay Details
| Gene Symbol: krt1-c5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076654.1 | NP_001070122.1 | 6 - 7 | 1046 | 97 | |
| XM_009301953.3 | XP_009300228.1 | 6 - 7 | 1075 | 97 | ||
| GenBank mRNA | BC124445.1 | - | 6 - 7 | 1046 | 97 | |
More Information
Set Membership: |
|

krt1-c5
Genomic Map