Assay Details
| Gene Symbol: tp53i11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077249.1 | NP_001070717.1 | 4 - 5 | 489 | 100 | |
| XM_021467658.1 | XP_021323333.1 | 4 - 5 | 498 | 100 | ||
| GenBank mRNA | BC124630.1 | - | 4 - 5 | 489 | 100 | |
| BC164984.1 | - | 2 - 3 | 227 | 100 | ||
More Information
Set Membership: |
|

tp53i11b
Genomic Map