Assay Details
| Gene Symbol: mrpl18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077330.1 | NP_001070798.1 | 2 - 3 | 399 | 63 | |
| XM_005157321.3 | XP_005157378.1 | 2 - 3 | 435 | 63 | ||
| GenBank mRNA | BC124672.1 | - | 2 - 3 | 399 | 63 | |
More Information
Set Membership: |
|

mrpl18
Genomic Map