Assay Details
| Gene Symbol: crtc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077457.1 | NP_001070925.1 | 10 - 11 | 1220 | 81 | |
| GenBank mRNA | BC124409.1 | - | 10 - 11 | 1220 | 81 | |
| BC165399.1 | - | 10 - 11 | 1220 | 81 | ||
More Information
Set Membership: |
|

crtc1a
Genomic Map