Assay Details
| Gene Symbol: faah2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077462.1 | NP_001070930.1 | 6 - 7 | 917 | 140 | |
| GenBank mRNA | BC124392.1 | - | 6 - 7 | 917 | 140 | |
| BC165762.1 | - | 6 - 7 | 902 | 140 | ||
More Information
Set Membership: |
|

faah2b
Genomic Map