Assay Details
| Gene Symbol: cyp4v8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077602.1 | NP_001071070.1 | 5 - 6 | 680 | 74 | |
| GenBank mRNA | BC125968.1 | - | 5 - 6 | 680 | 74 | |
| BC165389.1 | - | 5 - 6 | 687 | 74 | ||
More Information
Set Membership: |
|

cyp4v8
Genomic Map