Assay Details
| Gene Symbol: cyp4v8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077602.1 | NP_001071070.1 | 6 - 7 | 801 | 83 | |
| GenBank mRNA | BC125968.1 | - | 6 - 7 | 801 | 83 | |
| BC165389.1 | - | 6 - 7 | 808 | 83 | ||
More Information
Set Membership: |
|

cyp4v8
Genomic Map