Assay Details
| Gene Symbol: slc16a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080028.1 | NP_001073497.1 | 4 - 5 | 1114 | 69 | |
| XM_005160300.4 | XP_005160357.1 | 4 - 5 | 1149 | 69 | ||
| GenBank mRNA | BC129246.1 | - | 4 - 5 | 1114 | 69 | |
More Information
Set Membership: |
|

slc16a10
Genomic Map