Assay Details
| Gene Symbol: col15a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021470422.1 | XP_021326097.1 | 2241 | 127 | ||
| GenBank mRNA | AM259122.1 | - | 2167 | 127 | ||
More Information
Set Membership: |
|

col15a1a
Genomic Map