Assay Details
| Gene Symbol: col15a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AM259122.1 | - | 1476 | 80 | ||
| BC168501.1 | - | 1552 | 80 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

col15a1a| Gene Symbol: col15a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AM259122.1 | - | 1476 | 80 | ||
| BC168501.1 | - | 1552 | 80 | |||
Set Membership: |
|