Assay Details
| Gene Symbol: ubn2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080169.1 | NP_001073638.1 | 4 - 5 | 892 | 129 | |
| XM_005165715.4 | XP_005165772.1 | 4 - 5 | 1501 | 129 | ||
| XM_017356493.2 | XP_017211982.1 | 3 - 4 | 1399 | 129 | ||
| GenBank mRNA | BC129172.1 | - | 4 - 5 | 892 | 129 | |
More Information
Set Membership: |
|

ubn2a
Genomic Map