Assay Details
| Gene Symbol: dtnbb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080573.1 | NP_001074042.1 | 12 - 13 | 1391 | 67 | |
| GenBank mRNA | DQ516345.1 | - | 12 - 13 | 1391 | 67 | |
More Information
Set Membership: |
|

dtnbb
Genomic Map