Assay Details
| Gene Symbol: dtnbb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080573.1 | NP_001074042.1 | 14 - 15 | 1644 | 89 | |
| XM_017351659.2 | XP_017207148.1 | 17 - 18 | 2140 | 89 | ||
| XM_021467184.1 | XP_021322859.1 | 18 - 19 | 2221 | 89 | ||
| XM_021467185.1 | XP_021322860.1 | 18 - 19 | 2221 | 89 | ||
| XM_021467186.1 | XP_021322861.1 | 18 - 19 | 2222 | 89 | ||
| XM_021467187.1 | XP_021322862.1 | 18 - 19 | 2221 | 89 | ||
| GenBank mRNA | DQ516345.1 | - | 14 - 15 | 1644 | 89 | |
More Information
Set Membership: |
|

dtnbb
Genomic Map