Assay Details
| Gene Symbol: jam3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082863.3 | NP_001076332.2 | 8 - 9 | 968 | 94 | |
| GenBank mRNA | BC134233.1 | - | 8 - 9 | 968 | 94 | |
| CU458938.1 | - | 8 - 9 | 891 | 94 | ||
More Information
Set Membership: |
|

jam3b
Genomic Map