Assay Details
| Gene Symbol: tmem120a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082983.1 | NP_001076452.1 | 10 - 11 | 933 | 81 | |
| GenBank mRNA | BC133884.1 | - | 10 - 11 | 933 | 81 | |
More Information
Set Membership: |
|

tmem120a
Genomic Map