Assay Details
| Gene Symbol: erich3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083018.2 | NP_001076487.2 | 6 - 7 | 759 | 91 | |
| XM_009303859.2 | XP_009302134.1 | 6 - 7 | 855 | 91 | ||
| XM_009303860.2 | XP_009302135.1 | 6 - 7 | 852 | 91 | ||
| XM_009303862.2 | XP_009302137.1 | 6 - 7 | 842 | 91 | ||
| GenBank mRNA | BC122163.1 | - | 6 - 7 | 759 | 91 | |
| BC133135.1 | - | 6 - 7 | 745 | 91 | ||
More Information
Set Membership: |
|

erich3
Genomic Map